missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene in Toluene 100μg/mL, Fisher Chemical™

Product Code. 12933034 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
12933034 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 12933034

Brand: Fisher Chemical P872790

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 95-80-7
Physical Form Liquid
Molecular Formula C7H10N2
Concentration 100 μg/mL
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC1=CC=C(N)C=C1N
IUPAC Name 4-methylbenzene-1,3-diamine
Solvent or Matrix Toluene
Molecular Weight (g/mol) 122.17
PubChem CID 7261
Quantity 5 mL
ChEBI CHEBI:34237
Chemical Name or Material 2,4-Diaminotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.