missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenol in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12993034 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12993034 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12993034 Supplier Fisher Chemical Supplier No. P872850

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Especificaciones

Packaging Ampoule
CAS 105-67-9
Physical Form Liquid
Molecular Formula C8H10O
Concentration 100 μg/mL
UN Number 1230
InChI Key KUFFULVDNCHOFZ-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC1=CC(=C(C=C1)O)C
IUPAC Name 2,4-dimethylphenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 122.167
PubChem CID 7771
Quantity 1 mL
ChEBI CHEBI:34241
Chemical Name or Material 2,4-Dimethylphenol
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Standard solution 2,4-xylenol 100ug/ml in methanol
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.