missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline in Acetonitrile 10μg/mL, Fisher Chemical™

Product Code. 12942424 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity unitSize
12942424 10 mL 10mL
1 options
This item is not returnable. View return policy

Product Code. 12942424

Brand: Fisher Chemical P810800

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 95-51-2
Molecular Formula C6H6ClN
Concentration 10 μg/mL
UN Number 1648
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N)Cl
IUPAC Name 2-chloroaniline
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 127.571
PubChem CID 7240
Quantity 10 mL
Chemical Name or Material 2-Chloroaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution 2-chloroaniline 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.