missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoic acid methyl ester in Methanol 1000μg/mL, Fisher Chemical™

Product Code. 12987093 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12987093 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12987093 Supplier Fisher Chemical Supplier No. P884190

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 2905-67-1
Analyte or Component Names 3,5-Dichlorobenzoic acid methyl ester
Molecular Formula C8H6Cl2O2
Concentration 1000 μg/mL
MDL Number MFCD00032710
InChI Key BTEVDFJXGLQUDS-UHFFFAOYSA-N
Type Mass Spectrometry Standard
SMILES COC(=O)C1=CC(Cl)=CC(Cl)=C1
Element 3,5-Dichlorobenzoic Acid Methyl Ester
IUPAC Name methyl 3,5-dichlorobenzoate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 205.03
PubChem CID 76192
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.