missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoic acid in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™

Product Code. 12916053 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12916053 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12916053 Supplier Fisher Chemical Supplier No. F114621

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 51-36-5
Molecular Formula C7H4Cl2O2
Concentration 1000 μg/mL
InChI Key CXKCZFDUOYMOOP-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=C(C=C(C=C1Cl)Cl)C(=O)O
IUPAC Name 3,5-dichlorobenzoic acid
Solvent or Matrix Methyl-tert.butylether
Molecular Weight (g/mol) 191.007
PubChem CID 5811
Quantity 1 mL
Chemical Name or Material 3,5-Dichlorobenzoic acid
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.