missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl in Ethyl Acetate 1000μg/mL, Fisher Chemical™

Product Code. 12996023 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12996023 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996023 Supplier Fisher Chemical Supplier No. F113651

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 92-86-4
Molecular Formula C12H8Br2
Concentration 1000 μg/mL
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
Solvent or Matrix Ethyl Acetate
Molecular Weight (g/mol) 312.00
PubChem CID 7110
Quantity 1 mL
Chemical Name or Material 4,4prime-Dibromobiphenyl (PBB 15)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.