missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline in Acetonitrile 10μg/mL, Fisher Chemical™

Produktkod. 12942434 Handla allt Fisher Chemical Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
10 mL
Förpackningsstorlek:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
12942434 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 12942434 Leverantör Fisher Chemical Leverantörsnummer P810940

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifikationer

Packaging Ampoule(s)
CAS 106-47-8
Molecular Formula C6H6ClN
Concentration 10 μg/mL
UN Number 1648
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)Cl
IUPAC Name 4-chloroaniline
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 127.571
PubChem CID 7812
Quantity 10 mL
ChEBI CHEBI:20331
Chemical Name or Material 4-Chloroaniline
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • Standard solution 4-Chloroaniline 10ug/ml in acetonitrile
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.