missing translation for 'onlineSavingsMsg'
Learn More

Metabolite 4-Chloroaniline in DCM 100μg/mL, Fisher Chemical™

Product Code. 12992434 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12992434

Brand: Fisher Chemical P810990

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 106-47-8
Molecular Formula C6H6ClN
Concentration 100 μg/mL
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)Cl
IUPAC Name 4-chloroaniline
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 127.571
PubChem CID 7812
Quantity 1 mL
ChEBI CHEBI:20331
Chemical Name or Material 4-Chloroaniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.