missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12913024 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12913024 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12913024 Supplier Fisher Chemical Supplier No. P872670

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 103-33-3
Physical Form Liquid
Molecular Formula C12H10N2
Concentration 100 μg/mL
MDL Number MFCD00003022
InChI Key DMLAVOWQYNRWNQ-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC=C(C=C1)N=NC1=CC=CC=C1
IUPAC Name diphenyldiazene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 182.23
PubChem CID 2272
Quantity 1 mL
ChEBI CHEBI:190358
Chemical Name or Material Azobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • 1 ML Metabolite Azobenzene in Methanol 100µg/mL
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
Kompletterande information
  • MIXTURE LIST-Contain: methanol
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.