missing translation for 'onlineSavingsMsg'
Learn More

Bifenox in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12973174 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12973174 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12973174 Supplier Fisher Chemical Supplier No. P874320
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 42576-02-3
Molecular Formula C14H9Cl2NO5
Concentration 100 μg/mL
MDL Number MFCD00055314
InChI Key SUSRORUBZHMPCO-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(OC2=CC=C(Cl)C=C2Cl)=CC=C1[N+]([O-])=O
IUPAC Name methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 342.13
PubChem CID 39230
Quantity 1 mL
ChEBI CHEBI:6613
Chemical Name or Material Bifenox
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.