missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Carbazole in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12989943 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12989943 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12989943 Supplier Fisher Chemical Supplier No. P832400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 86-74-8
Molecular Formula C12H9N
Concentration 100 μg/mL
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name 9H-carbazole
Solvent or Matrix Methanol
Molecular Weight (g/mol) 167.21
PubChem CID 6854
Quantity 1 mL
ChEBI CHEBI:27543
Chemical Name or Material Carbazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.