missing translation for 'onlineSavingsMsg'
Learn More

Chlormequat Chloride in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12902424 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12902424 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12902424 Supplier Fisher Chemical Supplier No. P810670
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 999-81-5
Molecular Formula C5H13Cl2N
Concentration 100 μg/mL
UN Number 1648
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
SMILES C[N+](C)(C)CCCl.[Cl-]
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 158.066
PubChem CID 13836
Quantity 1 mL
Chemical Name or Material Chlormequat Chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chlormequat chloride,2-chloroethyltrimethylammonium chloride 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.