missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Chlorthal-diacid in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12972614 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12972614

Brand: Fisher Chemical P815360

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 1861-32-1
Molecular Formula C10H6Cl4O4
Concentration 100 μg/mL
MDL Number MFCD00014902
UN Number 1145
InChI Key NPOJQCVWMSKXDN-UHFFFAOYSA-N
SMILES COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl
IUPAC Name 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 331.95
PubChem CID 2943
Quantity 1 mL
ChEBI CHEBI:34664
Chemical Name or Material Chlorthal-diacid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chlorthal-diacid 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.