missing translation for 'onlineSavingsMsg'
Learn More

Dibromochloromethane in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12975304 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12975304 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12975304 Supplier Fisher Chemical Supplier No. P817230
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 124-48-1
Molecular Formula CHBr2Cl
Concentration 100 μg/mL
InChI Key GATVIKZLVQHOMN-UHFFFAOYSA-N
SMILES C(Cl)(Br)Br
IUPAC Name dibromo(chloro)methane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 208.277
PubChem CID 31296
Quantity 1 mL
ChEBI CHEBI:34627
Chemical Name or Material Dibromochloromethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1 ML Metabolite Dibromochloromethane in Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dibromochloromethane, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.