missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Dichlorvos D6 in n-Nonane 100μg/mL, Fisher Chemical™

Product Code. 12915454 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12915454

Brand: Fisher Chemical P819940

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 62-73-7
Molecular Formula C4H7Cl2O4P
Concentration 100 μg/mL
MDL Number MFCD00036123
InChI Key OEBRKCOSUFCWJD-UHFFFAOYSA-N
SMILES COP(=O)(OC)OC=C(Cl)Cl
IUPAC Name 2,2-dichloroethenyl dimethyl phosphate
Solvent or Matrix n-Nonane
Molecular Weight (g/mol) 220.97
PubChem CID 3039
Quantity 1 mL
ChEBI CHEBI:34690
Chemical Name or Material Dichlorvos D6
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.