missing translation for 'onlineSavingsMsg'
Learn More

Diquat dibromide hydrate in Water 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12936593 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12936593 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12936593 Supplier Fisher Chemical Supplier No. P822650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 6385-62-2
Molecular Formula C12H14Br2N2O
Concentration 100 μg/mL
MDL Number MFCD00211319
InChI Key KLJOSQAQZMLLMB-UHFFFAOYSA-L
SMILES O.[Br-].[Br-].C1C[N+]2=CC=CC=C2C2=CC=CC=[N+]12
IUPAC Name 7λ⁵,10λ⁵-diazatricyclo[8.4.0.0²,⁷]tetradeca-1(14),2,4,6,10,12-hexaene-7,10-bis(ylium) hydrate dibromide
Solvent or Matrix Water
Molecular Weight (g/mol) 362.07
PubChem CID 115268
Quantity 1 mL
Chemical Name or Material Diquat dibromide hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.