missing translation for 'onlineSavingsMsg'
Learn More

Gluphosinate ammonium in water 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12905994 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12905994 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12905994 Supplier Fisher Chemical Supplier No. P876460

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 77182-82-2
Molecular Formula C5H15N2O4P
Concentration 100mu;g/mL
MDL Number MFCD00055562,MFCD00211362
InChI Key ZBMRKNMTMPPMMK-UHFFFAOYNA-N
SMILES [NH4+].CP(O)(=O)CCC(N)C([O-])=O
IUPAC Name ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate
Solvent or Matrix Water
Molecular Weight (g/mol) 198.16
PubChem CID 122130249
Quantity 1 mL
Chemical Name or Material Gluphosinate ammonium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.