missing translation for 'onlineSavingsMsg'
Learn More

Heptachlor-exo-epoxide in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12936853 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12936853 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12936853 Supplier Fisher Chemical Supplier No. P828000
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 1024-57-3
Molecular Formula C10H5Cl7O
Concentration 100mu;g/mL
InChI Key ZXFXBSWRVIQKOD-GOWUZIGHSA-N
SMILES C12C(C(C3C1O3)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl
Solvent or Matrix Methanol
Molecular Weight (g/mol) 389.299
PubChem CID 94865622
Quantity 1 mL
Chemical Name or Material Heptachlor-exo-epoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.