missing translation for 'onlineSavingsMsg'
Learn More

Metamitron in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12989903 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12989903 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12989903 Supplier Fisher Chemical Supplier No. P831410
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifica

Packaging Ampoule(s)
CAS 41394-05-2
Molecular Formula C10H10N4O
Concentration 100 μg/mL
InChI Key VHCNQEUWZYOAEV-UHFFFAOYSA-N
SMILES CC1=NN=C(C(=O)N1N)C2=CC=CC=C2
IUPAC Name 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one
Solvent or Matrix Methanol
Molecular Weight (g/mol) 202.217
PubChem CID 38854
Quantity 1 mL
ChEBI CHEBI:6791
Chemical Name or Material Metamitron
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • 1 ML Metabolite Metamitron in Methanol 100µg/mL
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contain: methanol
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.