missing translation for 'onlineSavingsMsg'
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl) hydroxyl-amine hydrochloride in Water 15μg/mL, Fisher Chemical™

Product Code. 12976053 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12976053 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12976053 Supplier Fisher Chemical Supplier No. F114821

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 57981-02-9
Molecular Formula C7H5ClF5NO
Concentration 15 μg/mL
MDL Number MFCD00012953
InChI Key HVMVKNXIMUCYJA-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F
IUPAC Name hydrogen O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine chloride
Solvent or Matrix Water
Molecular Weight (g/mol) 249.57
PubChem CID 122307
Quantity 10 mL
Chemical Name or Material PFBHA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.