missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oryzalin in Methanol 1000μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 19044-88-3 |
| Molecular Formula | C12H18N4O6S |
| Concentration | 1000 μg/mL |
| InChI Key | UNAHYJYOSSSJHH-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])S(=O)(=O)N)[N+](=O)[O-] |
| IUPAC Name | 4-(dipropylamino)-3,5-dinitrobenzenesulfonamide |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 346.358 |
| Show More |
Safety and Handling
Product Identifier
- NCC Standard Solution - Oryzalin 1000ug/mL in Methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- EUH208-Contains . May produce an allergic reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands, forearms andface thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction