missing translation for 'onlineSavingsMsg'
Learn More

Phosphonic acid in Water 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12989503 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12989503 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12989503 Supplier Fisher Chemical Supplier No. P803900
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 1066-51-9
Physical Form Liquid
Molecular Formula CH6NO3P
Concentration 100 μg/mL
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C(N)P(=O)(O)O
IUPAC Name aminomethylphosphonic acid
Solvent or Matrix Water
Molecular Weight (g/mol) 111.037
PubChem CID 14017
Quantity 1 mL
ChEBI CHEBI:28812
Chemical Name or Material Phosphonic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.