missing translation for 'onlineSavingsMsg'
Learn More

Pyrene in DCM 10μg/mL, Fisher Chemical™

Product Code. 12996503 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12996503 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996503 Supplier Fisher Chemical Supplier No. P850290

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 129-00-0
Physical Form Liquid
Molecular Formula C16H10
Concentration 10 μg/mL
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
IUPAC Name pyrene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 202.256
PubChem CID 31423
Quantity 10 mL
ChEBI CHEBI:39106
Chemical Name or Material Pyrene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.