missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethene in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12975384 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12975384 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12975384 Supplier Fisher Chemical Supplier No. P818750

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 156-60-5
Molecular Formula C2H2Cl2
Concentration 10 μg/mL
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
SMILES Cl\C=C\Cl
IUPAC Name (1E)-1,2-dichloroethene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 96.94
PubChem CID 638186
Quantity 10 mL
ChEBI CHEBI:29027
Chemical Name or Material trans-1,2-Dichloroethene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.