missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphate in Ethyl Acetate 100μg/mL, Fisher Chemical™

Product Code. 12910354 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12910354 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12910354 Supplier Fisher Chemical Supplier No. P857260

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 115-86-6
Physical Form Liquid
Molecular Formula C18H15O4P
Concentration 100 μg/mL
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
IUPAC Name triphenyl phosphate
Solvent or Matrix Ethyl Acetate
Molecular Weight (g/mol) 326.29
PubChem CID 8289
Quantity 1 mL
ChEBI CHEBI:35033
Chemical Name or Material Triphenylphosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1 ML Metabolite Triphenylphosphate in Ethyl Acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl Acetate, Triphenyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.