missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphate in Methyl-tert.butylether 100μg/mL, Fisher Chemical™

Product Code. 12990344 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12990344 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12990344 Supplier Fisher Chemical Supplier No. P857240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 115-86-6
Physical Form Liquid
Molecular Formula C18H15O4P
Concentration 100 μg/mL
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
IUPAC Name triphenyl phosphate
Solvent or Matrix Methyl-tert.butylether
Molecular Weight (g/mol) 326.29
PubChem CID 8289
Quantity 1 mL
ChEBI CHEBI:35033
Chemical Name or Material Triphenylphosphate
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.