missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metadherin Antibody (CL0397) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52929AF647
This item is not returnable.
View return policy
Description
Metadherin Monoclonal antibody specifically detects Metadherin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Metadherin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC | |
| This antibody was developed using a recombinant protein derived from Q86UE4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Phospho Specific | |
| 92140 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0397 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction