missing translation for 'onlineSavingsMsg'
Learn More

Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 5000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17993726
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
Unit Size:
100g
200g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993726 100 g 100g
17911076 200 g 200g
17961066 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993726 Supplier VHG Labs Supplier No. VHGMA55000100G
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 5000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Calcium/Magnesium/Phosphorus/Zinc
CAS 7440-39-3, 7440-70-2, 7439-95-4, 7723-14-0, 7440-66-6
Methods ICP/XRF/RDE
Analyte or Component Names Barium/Calcium/Magnesium/Phosphorus/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Hydrocarbon Oil
Quantity 100 g
Concentration or Composition (by Analyte or Components) Barium 5000 μg/g/Calcium 5000 μg/g/Magnesium 5000 μg/g/Phosphorus 5000 μg/g/Zinc 5000 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.