Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-480
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
Brand:  Brady™ 104186
 View more versions of this product
Product Code. 11549433
  • ONLINE EXCLUSIVE PRICE
    992.00 €
    957.66 € / 1 rolls
    Save 34.34 €
    3% Off
In Stock
Add to basket
Description
Description
  • Available in various sizes
  • Rubber-based Mondo Bondo™ adhesive for oily, greasy applications
Specifications
Specifications
UL Recognised; cUL Recognised
Permanent Rubber
Light Gray
Roll
Single-side Printable
Label
Asset Tracking, General and Industrial Identificationentification, Rating Plates
41.91 mm
-70°C
10000 Label(s) / Roll
Rectangle
38.1 x 19.05 mm (W x H)
-70°C to +110°C
0.112 mm
19.05 x 38.10 mm
1.5 in.
38.10 mm
19.05 mm
Thermal Transfer Printable Labels
None
Satin
Polyester (B-480)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
2
THT-6-480-10
10,000/Pk.
38.10 mm (W) x 19.05 mm (H)
-94°F to +230°F
0.0044 in.
22.23 mm
0.75 in.
38.1 mm
0.75 in.
THT-6-480-10