Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-480
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
329.60 € - 1063.56 €
Specifications
Adhesive Type Permanent Rubber
Coating None
Color Light Gray
Finish Satin
Format Roll
View More Specs
Products 13
Product Code Brand Size (Metric) Price Quantity & Availability  
Product Code Brand Size (Metric) Price Quantity & Availability  
11539443
View Documents
Brady™
104196
W x H: 50.8 mm x 91.4m
ONLINE EXCLUSIVE PRICE
539.00 €
520.92 € / 1 rolls
Save 18.08 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11549443
View Documents
Brady™
104197
W x H: 101.6 mm x 91.4m
ONLINE EXCLUSIVE PRICE
1105.00 €
1063.56 € / 1 rolls
Save 41.44 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11519433
View Documents
Brady™
104183
19.05 x 6.35 mm (W x H)
ONLINE EXCLUSIVE PRICE
445.00 €
430.81 € / 1 rolls
Save 14.19 €
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
11579433
View Documents
Brady™
104190
50.8 x 25.4 mm (W x H)
ONLINE EXCLUSIVE PRICE
371.00 €
358.75 € / 1 rolls
Save 12.25 €
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
11599433
View Documents
Brady™
104192
W x H: 76.2 mm x 25.4 mm
ONLINE EXCLUSIVE PRICE
529.00 €
508.88 € / 1 rolls
Save 20.12 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11509443
View Documents
Brady™
104193
W x H: 76.2 mm x 50.8 mm
ONLINE EXCLUSIVE PRICE
351.00 €
339.18 € / 1 rolls
Save 11.82 €
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
11529443
View Documents
Brady™
104195
W x H: 76.2 mm x 91.4m
ONLINE EXCLUSIVE PRICE
750.00 €
724.07 € / 1 rolls
Save 25.93 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11529433
View Documents
Brady™
104184
25.4 x 9.52 mm (W x H)
ONLINE EXCLUSIVE PRICE
529.00 €
510.63 € / 1 rolls
Save 18.37 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11559433
View Documents
Brady™
104187
38.1 x 12.7 mm (W x H)
ONLINE EXCLUSIVE PRICE
947.00 €
912.08 € / 1 rolls
Save 34.92 €
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
11569433
View Documents
Brady™
104189
50.8 mm x 12.7 mm (W x H)
ONLINE EXCLUSIVE PRICE
342.00 €
329.60 € / 1 rolls
Save 12.40 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11519443
View Documents
Brady™
104194
W x H: 101.6 mm x 50.8 mm
ONLINE EXCLUSIVE PRICE
458.00 €
440.79 € / 1 rolls
Save 17.21 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11549433
View Documents
Brady™
104186
38.1 x 19.05 mm (W x H)
ONLINE EXCLUSIVE PRICE
992.00 €
957.66 € / 1 rolls
Save 34.34 €
3% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
11589433
View Documents
Brady™
104191
W x H: 69.85 mm x 31.75 mm
ONLINE EXCLUSIVE PRICE
604.00 €
578.43 € / 1 rolls
Save 25.57 €
4% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
Description
Description
  • Available in various sizes
  • Rubber-based Mondo Bondo™ adhesive for oily, greasy applications
Specifications
Specifications
Permanent Rubber
Light Gray
Roll
Non-sterile
-70°C
-94°F to +230°F
0.0044 in.
None
Satin
Thermal Transfer
Asset Tracking, General and Industrial Identificationentification, Rating Plates
Rectangle
-70°C to +110°C
0.112 mm