Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-480
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
Brand:  Brady™ 104189
 View more versions of this product
Product Code. 11569433
  • ONLINE EXCLUSIVE PRICE
    342.00 €
    329.60 € / 1 rolls
    Save 12.40 €
    3% Off
In Stock
Add to basket
Description
Description
  • Available in various sizes
  • Rubber-based Mondo Bondo™ adhesive for oily, greasy applications
Specifications
Specifications
UL Recognised; cUL Recognised
Permanent Rubber
Light Gray
Roll
Single-side Printable
Label
Asset Tracking, General and Industrial Identificationentification, Rating Plates
1
THT-53-480-3
3000/Pk.
50.80 mm (W) x 12.70 mm (H)
-94°F to +230°F
0.0044 in.
15.24 mm
0.5 in.
50.8 mm
0.5 in.
THT-53-480-3
Thermal Transfer Printable Labels
None
Satin
Polyester (B-480)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
-70°C
3000 Label(s) / Roll
Rectangle
50.8 mm x 12.7 mm (W x H)
-70°C to +110°C
0.112 mm
12.70 x 50.80 mm
2 in.
50.80 mm
12.7 mm