missing translation for 'onlineSavingsMsg'
Learn More

Metalaxyl metabolit CGA 108906, TRC

Product Code. 30180229
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180229 50 mg 50mg
30179756 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180229 Supplier Toronto Research Chemicals Supplier No. TRCM26081050MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104390-56-9
Molecular Formula C14 H17 N O6
Molecular Weight (g/mol) 295.29
Synonym 2-[(1-Carboxyethyl)(2-methoxyacetyl)amino]-3-methylbenzoic acid, 2-[(1-Carboxyethyl)(methoxyacetyl)amino]-3-methyl-benzoic Acid, Benzoic acid, 2-[(1-carboxyethyl)(methoxyacetyl)amino]-3-methyl- (9CI), Benzoic acid, 2-[(1-carboxyethyl)(methoxyacetyl)amino]-3-methyl-, (±)-, Metalaxyl metabolite CGA 108906
IUPAC Name 2-[1-carboxyethyl-(2-methoxyacetyl)amino]-3-methylbenzoic acid
SMILES COCC(=O)N(C(C)C(=O)O)c1c(C)cccc1C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H17NO6/c1-8-5-4-6-10(14(19)20)12(8)15(9(2)13(17)18)11(16)7-21-3/h4-6,9H,7H2,1-3H3,(H,17,18)(H,19,20)
Formula Weight 295.1056
Chemical Name or Material 2-[(1-carboxyethyl)(methoxyacetyl)amino]-3-methyl-benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.