missing translation for 'onlineSavingsMsg'
Learn More

Metals in Biodiesel Standard: Ca, K, Mg, Na, P @ 5 μg/g in B100 Biodiesel, VHG™

Product Code. 17919974
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919974 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919974 Supplier VHG Labs Supplier No. VHGB100M55100G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Metals in Biodiesel Standard: Ca, K, Mg, Na, P @ 5 μg/g in B100 Biodiesel
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Magnesium/Phosphorus/Potassium/Sodium
CAS 7440-70-2, 7439-95-4, 7723-14-0, 9-7-7440, 7440-23-5
Analyte or Component Names Calcium/Magnesium/Phosphorus/Potassium/Sodium
Shipping Condition Room Temperature
Solvent or Matrix B100 Biodiesel
Quantity 100 g
Concentration or Composition (by Analyte or Components) Calcium 5 μg/g/Magnesium 5 μg/g/Phosphorus 5 μg/g/Potassium 5 μg/g/Sodium 5 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.