missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Phosphoprotein Antibody (hMPV33) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50506-0.025ml
This item is not returnable.
View return policy
Description
Metapneumovirus Phosphoprotein Monoclonal antibody specifically detects Metapneumovirus Phosphoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Metapneumovirus Phosphoprotein | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunofluorescence | |
| hMPV33 | |
| Unconjugated | |
| PBS | |
| Human hMPV strain NCL-174 | |
| 0.025 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction