missing translation for 'onlineSavingsMsg'
Learn More

Metaraminol Bitartrate, TRC

Product Code. 30477908
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477908 500 mg 500mg
30475938 100 mg 100mg
30472874 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477908 Supplier Toronto Research Chemicals Supplier No. TRCM225565500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33402-03-8
Molecular Formula C9 H13 N O2 . C4 H6 O6
Molecular Weight (g/mol) 317.29
Synonym Benzenemethanol, α-[(1S)-1-aminoethyl]-3-hydroxy-, (αR)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Benzenemethanol, α-(1-aminoethyl)-3-hydroxy-, [R-(R*,S*)]-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Benzenemethanol, α-[(1S)-1-aminoethyl]-3-hydroxy-, (αR)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Benzyl alcohol, α-(1-aminoethyl)-m-hydroxy-, (-)-, tartrate (1:1) (salt) (8CI), (-)-Metaraminol bitartrate, Aramine, Aramine (sympathomimetic), Icoral B, Metaradrine bitartrate, Metaraminol bitartrate, Metaraminol hydrogen tartrate, Metaraminol tartrate, Pressorol
IUPAC Name 3-[(1R,2S)-2-amino-1-hydroxypropyl]phenol;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES C[C@H](N)[C@H](O)c1cccc(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H13NO2.C4H6O6/c1-6(10)9(12)7-3-2-4-8(11)5-7;5-1(3(7)8)2(6)4(9)10/h2-6,9,11-12H,10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t6-,9-;1-,2-/m01/s1
Formula Weight 317.1111
Chemical Name or Material Metaraminol Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.