missing translation for 'onlineSavingsMsg'
Learn More

Metformin hydrochloride, MedChemExpress

Product Code. 16477358
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477358 10 g 10g
16413387 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477358 Supplier Medchem Express Supplier No. HY17471A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Metformin hydrochloride (1,1-Dimethylbiguanide hydrochloride) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy.

Metformin hydrochloride (1,1-Dimethylbiguanide hydrochloride) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy.

Specifications

Chemical Name or Material Metformin hydrochloride
Physical Form Solid
CAS 1115-70-4
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C4H12ClN5
For Use With (Application) Cancer-Kinase/protease
Synonym 1,1-Dimethylbiguanide hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (603.79 mM) ∣DMSO : ≥ 1.7 mg/mL (10.26 mM)
SMILES NC(NC(N(C)C)=N)=N.Cl
Molecular Weight (g/mol) 165.62
Formula Weight 165.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.