missing translation for 'onlineSavingsMsg'
Learn More

Methacholine chloride, MedChemExpress

Product Code. 16438026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438026 Supplier Medchem Express Supplier No. HYA0083
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methacholine (Acetyl-β-methylcholine) choride is a potent muscarinic-3 (M3) agonist.

Methacholine choride acts directly on acetylcholine receptors on smooth muscle causing bronchoconstriction and airway narrowing. Methacholine choride shows a high sensitivity to identify bronchial hyperresponsiveness (BHR). Methacholine choride can be used to measure airway hyperresponsiveness (AHR) as a diagnostic aid in the assessment of individuals with asthma-like symptoms and normal resting expiratory flow rates.

Specifications

Chemical Name or Material Methacholine chloride
Physical Form Solid
CAS 62-51-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H18ClNO2
Synonym Acetyl-β-methylcholine chloride
Solubility Information H2O : ≥ 100 mg/mL (511.01 mM) ∣DMSO : ≥ 42 mg/mL (214.63 mM)
SMILES CC(OC(C)=O)C[N+](C)(C)C.[Cl-]
Molecular Weight (g/mol) 195.69
Formula Weight 195.69
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.