missing translation for 'onlineSavingsMsg'
Learn More

Methacrolein, 90%, stabilized

Product Code. 10267073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10267073 10 mL Glass bottle 10mL
10275253 50 mL Glass bottle 50mL
10069560 100 mL Glass bottle 100mL
10643572 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10267073 Supplier Thermo Scientific Acros Supplier No. 292830100
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

Methacrolein, 90%, C4H6O, CAS Number-78-85-3, 2-methyl-2-propenal, 2-methylpropenal, 2-methylacrolein, 2-methylenepropanal, 2-propenal, 2-methyl, isobutenal, methacrylaldehyde, methacrylic aldehyde, methylacrylaldehyde, methacrolein, 50mL, 88% min. (GC), 01, III, 2981, 69.0 deg.C | CAS: 78-85-3 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-85-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00006974
InChI Key STNJBCKSHOAVAJ-UHFFFAOYSA-N
Synonym methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
PubChem CID 6562
IUPAC Name 2-methylprop-2-enal
SMILES CC(=C)C=O

Specifications

Melting Point -81.0°C
Color Colorless to Yellow
Density 0.8470g/mL
Boiling Point 69.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CHO
Refractive Index 1.4145
Quantity 10 mL
Beilstein 01, III, 2981
Specific Gravity 0.847
Solubility Information Solubility in water: 6g/100mL (20°C).
Stabilizer 0.1% hydroquinone
Viscosity 0.49 mPa.s (20°C)
Formula Weight 70.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methacrolein, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.