missing translation for 'onlineSavingsMsg'
Learn More

Methallyl phenyl ether, 96%

Product Code. 11410467
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410467 5 g 5g
11420467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410467 Supplier Thermo Scientific Alfa Aesar Supplier No. B22369.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5820-22-4 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5820-22-4
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008597
InChI Key LECDNXOCIPRJNJ-UHFFFAOYSA-N
Synonym methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w
PubChem CID 79893
IUPAC Name 2-methylprop-2-enoxybenzene
SMILES CC(=C)COC1=CC=CC=C1

Specifications

Density 0.964
Boiling Point 204°C
Flash Point 77°C (171°F)
Refractive Index 1.515
Quantity 5 g
Beilstein 1933868
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material Methallyl phenyl ether
compliance-icons
Product Identifier
  • Methallyl phenyl ether
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: (2-Methylallyloxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.