missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methfuroxam, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Methfuroxam |
| CAS | 28730-17-8 |
| Analyte or Component Names | Methfuroxam |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H15 N O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-Furancarboxamide, 2,4,5-trimethyl-N-phenyl- (9CI, ACI), 2,4,5-Trimethyl-N-phenyl-3-furancarboxamide (ACI), 3-Furanilide, 2,4,5-trimethyl- (8CI), 2,4,5-Trimethyl-3-furanilide, Furavax, H 719, Methfuroxam, Trivax, UBI 2238, UBI-H 719 |
| InChI Formula | InChI=1S/C14H15NO2/c1-9-10(2)17-11(3)13(9)14(16)15-12-7-5-4-6-8-12/h4-8H,1-3H3,(H,15,16) |
| Solution Type | Neat |
| SMILES | Cc1oc(C)c(C(=O)Nc2ccccc2)c1C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction