missing translation for 'onlineSavingsMsg'
Learn More

Melford Methicillin Sodium Salt

Product Code. 30018491
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 30018491

Brand: Melford M628001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methicillin Sodium Salt is a chemical compound that belongs to the class of beta-lactam antibiotics known as penicillin.

Methicillin Sodium Salt is a chemical compound that belongs to the class of beta-lactam antibiotics known as penicillin.

It is a modified form of penicillin that was developed to be resistant to the effects of the enzyme penicillinase, which is produced by certain bacteria. This resistance allows methicillin to be effective against bacteria that have developed resistance to traditional penicillin antibiotics.

This chemical is primarily used as a selective agent in laboratory experiments to isolate and study bacteria that are resistant to methicillin and other related antibiotics. It is commonly used in microbiology research, particularly in studies focused on antibiotic resistance mechanisms and bacterial genetics.

Researchers may utilize Methicillin Sodium Salt in various techniques such as bacterial culture, antibiotic susceptibility testing, and identification of antibiotic resistance genes. Its use helps to understand the mechanisms of bacterial resistance and develop strategies to combat antibiotic-resistant pathogens.

Specifications

Chemical Name or Material Methicillin
CAS 132-92-3
Quantity 1 g
Molecular Formula C17H19N2O6SNa-H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.