missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methidathion 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Methidathion |
| CAS | 950-37-8 |
| Analyte or Component Names | Methidathion |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H11 N2 O4 P S3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Phosphorodithioic acid, S-[(5-methoxy-2-oxo-1,3,4-thiadiazol-3(2H)-yl)methyl] O,O-dimethyl ester, Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one (7CI,8CI), DMTP, DMTP (insecticide), Dithiophosphoric acid O,O'-dimethyl-S-[(2-methoxy-1,3,4-thiadiazol-5(4H)-on-4-yl)methyl] ester, Dithiophosphoric acid O,O'-dimethyl-S-[(5-methoxy-1,3,4-thiadiazol-2(3H)-on-3-yl)methyl] ester, GS 13005, Geigy 13005, Geigy GS 13005, Methidathion, Methidation, O,O-Dimethyl S-(5-methoxy-1,3,4-thiadiazole-2 (3H)-on-3-ylmethyl) dithiophosphate, O,O-Dimethyl S-[2-methoxy-1,3,4-thiadiazol-5(4H)-on-4-ylmethyl] phosphorodithioate, O,O-Dimethyl phosphorodithioate S-ester with 4-(mercaptomethyl)-2-methoxy-1,3,4-thiadizaolin-5-one, O,O-Dimethyl-S-(2-methoxy-1,3,4-thiadiazol-5(4H)-onyl-4-methyl) dithiophosphate, O,O-Dimethyl-S-(2-methoxy-Δ2-1,3,4-thiadiazolin-5-on-4-ylmethyl)dithiophosphate, O,O-Dimethyl-S-(5-methoxy-1,3,4-thiadiazol-2(3H)-on-3-ylmethyl)dithiophosphate, O,O-Dimethyl-S-2-methyloxy-1,3,4-thiodiazol-5-(4H)-onyl-(4)-methyldithiophosphate, O,O-Dimethyl-S-[2-methoxy-1,3,4-thiadiazol-5(4H)-onylmethyl] dithiophosphate, O,O-Dimethylphosphorodithioate S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one, O,O'-Dimethyl-S-[(2-methoxy-1,3,4-thiadiazole-5(4H)-one-4-yl)methyl]dithiophosphate, Phosphorodithioic acid O,O'-dimethyl ester S-ester with 4-(mercaptomethyl)-2-methoxy-Δ2-1,3,4-thiadiazolin-5-one, S-((2-Methoxy-5-oxo-Δ2-1,3,4-thiadiazolin-4-yl)methyl) O,O-dimethylphosphorodithioate, Shapulin, Supracid, Supracide, Suprathion, Ultracid, Ultracid 40, Ultracid EC 40 |
| InChI Formula | InChI=1S/C6H11N2O4PS3/c1-10-5-7-8(6(9)16-5)4-15-13(14,11-2)12-3/h4H2,1-3H3 |
| Solution Type | Single Solution |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?