Learn More
MP Biomedicals™ Methionine/Choline-deficient Diet, Pellets
Methionine/Choline-deficient Pellet Diet, is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.
Brand: MP Biomedicals™ 0296043920
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Non-alcoholic steatohepatitis(NASH) was induced by feeding a methionine/choline-deficient diet (MCD-diet) for up to 5 weeks to mice.
- NASH is a disease with a high incidence, difficult diagnosis, and as yet no effective treatment.
- So, MCD-diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.
Specifications
Choline Deficient Diet | |
Pellets | |
20 kg | |
Store at Room Temperature (15°C to 30°C) |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.