missing translation for 'onlineSavingsMsg'
Learn More

Method 8270B - Organophosphorous Pesticide Mix, 9-2 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30319199
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319199 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319199 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000364DI
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethoate∣Disulfoton∣Famphur∣O,O,O-Triethylphosphorothioate∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Sulfotep∣Thionazin
CAS 60-51-5, 298-04-4, 52-85-7, 126-68-1, 56-38-2, 298-00-0
Analyte or Component Names Dimethoate∣Disulfoton∣Famphur∣O,O,O-Triethylphosphorothioate∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Sulfotep∣Thionazin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Dimethoate 2000 μg/mL∣Disulfoton 2000 μg/mL∣Famphur 2000 μg/mL∣O,O,O-Triethylphosphorothioate 2000 μg/mL∣Parathion-ethyl 2000 μg/mL∣Parathion-methyl 2000 μg/mL∣Phorate 2000 μg/mL∣Sulfotep 2000 μg/mL∣Thionazin 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.