missing translation for 'onlineSavingsMsg'
Learn More

Methoxychlor, SPEX CertiPrep™

Product Code. 12939284 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12939284 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12939284 Supplier SPEX Certiprep Supplier No. S2397
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Methoxychlor72-43-51000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL Amber Prescored Glass Ampule
CAS 72-43-5, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C16H15Cl3O2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
Type Certified Reference Material
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 345.644
PubChem CID 4115
Quantity 1 mL
ChEBI CHEBI:6842
Chemical Name or Material Methoxychlor
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methoxychlor
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.