missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%

Product Code. 11461510
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461510 25 g 25g
11471510 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461510 Supplier Thermo Scientific Alfa Aesar Supplier No. H26930.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3939-01-3 | C7H3F2N | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3939-01-3
MDL Number MFCD00012799

Specifications

Melting Point ∽185°C (decomposition)
Molecular Formula C14H18ClNO3
Quantity 25 g
Sensitivity Hygroscopic
Synonym methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
InChI Key BRADBAOVPACOQQ-UHFFFAOYNA-N
SMILES COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
IUPAC Name methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
Molecular Weight (g/mol) 283.75
PubChem CID 107479
Formula Weight 283.75
Percent Purity 95%
Chemical Name or Material Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.