missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-C-[4-Chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-L-glucopyranoside, TRC

Product Code. 30478267
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478267 1 mg 1mg
30474003 50 mg 50mg
30482753 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478267 Supplier Toronto Research Chemicals Supplier No. TRCM3232881MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 1-C-[4-Chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-L-glucopyranoside
Analyte or Component Names Methyl 1-C-[4-Chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-L-glucopyranoside
Shipping Condition Room Temperature
Molecular Formula C22 H27 Cl O7
Synonym (3R,4S,5R,6S)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol
InChI Formula InChI=1S/C22H27ClO7/c1-3-29-16-7-4-13(5-8-16)10-14-11-15(6-9-17(14)23)22(28-2)21(27)20(26)19(25)18(12-24)30-22/h4-9,11,18-21,24-27H,3,10,12H2,1-2H3/t18-,19-,20+,21-,22?/m0/s1
Solution Type Neat
SMILES CCOc1ccc(Cc2cc(ccc2Cl)C3(OC)O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]3O)cc1
IUPAC Name (3S,4R,5R,6S)-2-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-6-(hydroxymethyl)-2-methoxyoxane-3,4,5-triol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 438.899
Quantity 1 mg
Formula Weight 438.145
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.