missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-hydroxy-2-naphthoate, Thermo Scientific Chemicals

Catalog Number 11925001
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
948-03-8
C12H10O3
202.21
HMIBDRSTVGFJPB-UHFFFAOYSA-N
methyl 1-hydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1-hydroxy-, methyl ester, 1-hydroxy-2-naphthoic acid methyl ester, 1-hydroxy-2-naphthoic acid, methyl ester, acmc-20akjf, 2-naphthoic acid, 1-hydroxy-, methyl ester, methyl=1-hydroxy-2-naphthoate, 1-hydroxy-2-carbomethoxynaphthalene, methyl 1-hydroxy-naphthalene-2-carboxylate
70358
methyl 1-hydroxynaphthalene-2-carboxylate
COC(=O)C1=C(C2=CC=CC=C2C=C1)O
This item is not returnable. View return policy
948-03-8
C12H10O3
202.21
HMIBDRSTVGFJPB-UHFFFAOYSA-N
methyl 1-hydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1-hydroxy-, methyl ester, 1-hydroxy-2-naphthoic acid methyl ester, 1-hydroxy-2-naphthoic acid, methyl ester, acmc-20akjf, 2-naphthoic acid, 1-hydroxy-, methyl ester, methyl=1-hydroxy-2-naphthoate, 1-hydroxy-2-carbomethoxynaphthalene, methyl 1-hydroxy-naphthalene-2-carboxylate
70358
methyl 1-hydroxynaphthalene-2-carboxylate
COC(=O)C1=C(C2=CC=CC=C2C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 80°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.21
Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
compliance-icons
Product Identifier
  • Methyl 1-hydroxy-2-naphthoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'