missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dihydroxybenzoate, 97%

Product Code. 11486466
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486466 25 g 25g
11476466 5 g 5g
11496466 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486466 Supplier Thermo Scientific Alfa Aesar Supplier No. B24757.14
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2150-47-2 | C8H8O4 | 168.148 g/mol

Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-47-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002276
InChI Key IIFCLXHRIYTHPV-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate
PubChem CID 16523
IUPAC Name methyl 2,4-dihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)O

Specifications

Melting Point 116°C to 119°C
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.