missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dihydroxybenzoate, 97%

Product Code. 10794733 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
10794733 5 g Glass bottle 5g
10275410 25 g Glass bottle 25g
10606524 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10794733

Brand: Thermo Scientific Acros 126730050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2150-47-2 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-47-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002276
InChI Key IIFCLXHRIYTHPV-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate
PubChem CID 16523
IUPAC Name methyl 2,4-dihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)O

Specifications

Melting Point 116°C to 121°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2CH3
Quantity 5 g
Beilstein 10,I,178
Formula Weight 168.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 2, 4-dihydroxybenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,4-dihydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.